HPLC columns, YMC-Pack™ Pro C18
Fournisseur: YMC
AS12S03-1546WT AS12S03-0546WT AS12S05-1503WT
YMCAAS12S03-1546WTEA 964 EUR InStock
YMCAAS12S03-1546WT YMCAAS12S03-0546WT YMCAAS12S05-1503WT
HPLC columns, YMC-Pack™ Pro C18
Colonnes pour chromatographie
The ProFamily range is manufactured from ultra high purity silica (99,999%), which is then subjected to a special bonding process. Silica with an extremely low metal content is used for the phases of the ProFamily to suppress polar interactions, as metal impurities influence silanol groups to become more acidic.
  • Advanced endcapping technology
  • Excellent resolution
  • Reproducibility
Ideal for the separation of acidic and basic molecules and analysis of of drugs or metabolites, compounds that are susceptible to polar interactions with residual silanol groups and metal impurities.
Information de livraison: Every column supplied with lot certificate on physicochemical properties of bare silica, minimal metal traces, bonding and chromatographic performance; test chromatogram showing high performance plate count together with tight specification on peak symmetry.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité