HPLC columns, YMC-Pack™ Hydrosphere Pro C18
Fournisseur: YMC
HS12S052546WT
YMCAHS12S052546WTEA 1074 EUR InStock
YMCAHS12S052546WT
HPLC columns, YMC-Pack™ Hydrosphere Pro C18
Colonnes pour chromatographie
The ProFamily range is manufactured from ultra high purity silica (99,999%), which is then subjected to a special bonding process. Silica with an extremely low metal content is used for the phases of the ProFamily to suppress polar interactions, as metal impurities influence silanol groups to become more acidic.
  • Advanced endcapping technology
  • Excellent resolution
  • Reproducibility
Ideal for the separation highly polar compounds.
Information de livraison: Every column supplied with lot certificate on physicochemical properties of bare silica, minimal metal traces, bonding and chromatographic performance; test chromatogram showing high performance plate count together with tight specification on peak symmetry.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité