Dissolved oxygen meters, handheld, EcoSense® DO200A
Fournisseur: YSI (ENVIRONMENTAL PRODUCTS
606040
YSIA606040EA 1160 EUR InStock
YSIA606040
Dissolved oxygen meters, handheld, EcoSense® DO200A
Electrochemistry Meters Oxygen Meters
The DO200A simultaneously measures dissolved oxygen (% air saturation and ppm) and temperature in wastewater, surface water, and aquaculture applications. The DO200A can also be used in the laboratory with the 200-BOD self-stirring lab probe making the DO200A a true lab and field instrument.
  • Calibrates easily in a saturated air environment and displays in % or ppm (mg/L) values
  • Screw on PE membranes make membrane changes easier than ever
  • Low battery indicator with 500 hour battery life
  • Automatic shutoff after 30 minutes of inactivity
  • Salinity, pressure and temperature compensation
Informations de commande: Kit includes meter, probe with cable (10 m), and hard-sided case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité