Zinc Formalin Fixative
Fournisseur: SIGMA-ALDRICH MICROSCOPY
Z2902-3.75L
Z2902-3.75LEA 140 EUR InStock
Z2902-3.75L
Zinc Formalin Fixative
Colorants Réactifs pour la microscopie et la coloration
Zinc formalin fixative is a non-precipitating fixative. This fixative is compatible with histochemical, immunochemical and special stains. It may be used with automated tissue processors and also in manual methods. The zinc chloride component of the formulation increases the rigidity of cellular components to withstand subsequent processing, sectioning and staining procedures. Tissue specimens initially fixed using 10% neutral buffered formalin solution may be post-fixed using the zinc formalin fixative.
Zinc formalin fixative is an excellent morphological preservation of nuclear and cytoplasmic components. Can replace neutral buffered formalin for routine tissue and immunohistochemical procedures. It is a non-precipitating fixative which can be used with automated and manual methods.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité