Image acquisition and processing under Windows 10 x64. User interface configurable, control of the ZEISS microscope systems and components, extensive acquisition and analysis. CZI image format. The following functions are included: - Apeer onsite adv 1 * 1 Pce
Fournisseur: ZEISS
COMMANDE
Développer tout / Réduire tout
Image acquisition and processing under Windows 10 x64. User interface configurable, control of the ZEISS microscope systems and components, extensive acquisition and analysis. CZI image format. The following functions are included: - Apeer onsite adv 1 * 1 Pce ZEIS4101350002370 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou