Digital camera, Axiocam 305 color
Fournisseur: ZEISS
4265609030000
ZEIS4265609030000EA 4400 EUR InStock
ZEIS4265609030000
Digital camera, Axiocam 305 color
Caméras
5 Megapixel microscope camera for routine and research labs. For use on compound, stereo, or zoom microscopes.
  • Fast readout with 36 images per second in full colour resolution
  • Highly sensitive CMOS sensor technology
  • Small 3,45 µm pixels for better sampling at low magnifications
  • Easy to use super-speed USB 3,0 connection
  • Colour and black and white imaging modes
  • Compatible with all Zeiss microscopes with camera port
  • Fast and efficient operation with ZEN imaging software
Recommended for: Applications with bright samples, documentation, routine tasks, materials research, quality assurance / quality control, fast high resolution live image for co-observation, fast image acquisition and time-lapse recording.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité