Axiocam 208 color Digital Camera
Fournisseur: ZEISS
4265709001000
ZEIS4265709001000EA 2580 EUR InStock
ZEIS4265709001000
Axiocam 208 color Digital Camera
Caméras
Axiocam 208 color is your smart versatile 8 megapixel color microscope camera suitable for education, documentation and routine applications. This CMOS camera delivers crisp, detail rich live images with high colour fidelity at full 4K resolution in outstanding 30 fps.
  • Full 4K resolution in outstanding 30 fps
  • Brilliant colour rendering
  • Live image enhancement functions like sharpening, denoising and HDR
  • Ethernet, USB 3.0
  • Wi-Fi compatible - use labscope imaging app to control your camera wirelessly
  • Document your samples as you see it in the eyepieces
Easy and effortless digital documentation - especially suitable for education, digital classroom and routine documentation. Use in stand-alone mode and save images on USB flash drive, use labscope imaging app or ZEN imaging software.
Stand-alone operation with camera control by intuitive on screen display via mouse and keyboard without a PC. Connect directly to a monitor by a HDMI cable for live image display for search and focussing and review of acquired images.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité