Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex®, L/S 35; 25 ft
Fournisseur: Avantor Fluid Handling
MFLX06424-35EA 171 EUR InStock
MFLX06424-35
Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex®, L/S 35; 25 ft
Tuyaux Tubes pour pompe péristaltique
VWR Item#MFLX06424-35
Your Organization's Price:
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
  • Tubing Size: L/S 35
Applications: General-purpose lab uses requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white
See 96123-15 for the irradiated version of this tubing.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
CE certificate 
Key Features
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
  • Tubing Size: L/S 35
More About This Item
See 96123-15 for the irradiated version of this tubing.
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité