Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, L/S 18; 25 ft
Fournisseur: Avantor Fluid Handling
MFLX06508-18EA 273 EUR InStock
MFLX06508-18
Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, L/S 18; 25 ft
Tuyaux Tubes pour pompe péristaltique
VWR Item#MFLX06508-18
Your Organization's Price:
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 18
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
See 96122-16 for the irradiated version of this tubing.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
CE certificate 
Key Features
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 18
More About This Item
See 96122-16 for the irradiated version of this tubing.
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité