Masterflex® L/S® Standard Pump Head with Long-Shaft for Precision Tubing, L/S® 16, Polycarbonate Housing, CRS Rotor
Fournisseur: Avantor Fluid Handling
MFLX07016-00EA 382 EUR InStock
MFLX07016-00
Masterflex® L/S® Standard Pump Head with Long-Shaft for Precision Tubing, L/S® 16, Polycarbonate Housing, CRS Rotor
Pompe Pump Heads Peristaltic Pump Heads
VWR Item#MFLX07016-00
Your Organization's Price:
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Suitable for use with L/S® 16 tubing size
  • Rotor is constructed of cold-rolled steel (CRS) for better chemical resistance
This Standard pump head accepts a single tubing size to ensure optimal performance from your Masterflex pump. Use with Masterflex L/S pump drives requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Suitable for use with L/S® 16 tubing size
  • Rotor is constructed of cold-rolled steel (CRS) for better chemical resistance
More About This Item
This Standard pump head accepts a single tubing size to ensure optimal performance from your Masterflex pump. Use with Masterflex L/S pump drives requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives.
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité