Masterflex® Extension Cable for L/S® and I/P® Precision Modular Drives; 9 ft
Fournisseur: Avantor Fluid Handling
MFLX07557-51EA 627 EUR InStock
MFLX07557-51
Masterflex® Extension Cable for L/S® and I/P® Precision Modular Drives; 9 ft
Câbles
VWR Item#MFLX07557-51
Your Organization's Price:
Increase modular drive flexibility with extended cable lengths.
  • Easily connect to your Masterflex® pump with connectors on both ends
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2,7 m) to increase installation flexibility. Do not extend more than 25 ft.
This extension cable is compatible with L/S® pump models 07557-00, -10, -60, and -70; and with I/P® pump models 07591-20 and -30.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Easily connect to your Masterflex® pump with connectors on both ends
More About This Item
This extension cable is compatible with L/S® pump models 07557-00, -10, -60, and -70; and with I/P® pump models 07591-20 and -30.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité