Masterflex® Cable Assembly for Remote Control, DB9(M) Connector; 25 Ft
Fournisseur: Avantor Fluid Handling
MFLX07595-47EA 369 EUR InStock
MFLX07595-47
Masterflex® Cable Assembly for Remote Control, DB9(M) Connector; 25 Ft
Câbles
VWR Item#MFLX07595-47
Your Organization's Price:
Easily connect your Masterflex® drive to a PLC or other control device.
  • Constructed of durable PVC
  • Long 25 ft (7.9 m) cable length makes connection simple
  • Cable features 26 AWG wire gauge
  • Connection Type: DB9(M)
Cable assemblies include terminal connector and 25 ft (7.9 m) cable with stripped wire ends. Verify the connector type on your drive before ordering
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Constructed of durable PVC
  • Long 25 ft (7.9 m) cable length makes connection simple
  • Cable features 26 AWG wire gauge
  • Connection Type: DB9(M)
More About This Item
Cable assemblies include terminal connector and 25 ft (7.9 m) cable with stripped wire ends. Verify the connector type on your drive before ordering
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité