Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Fournisseur: Avantor Fluid Handling
MFLX31200-90EA 171 EUR InStock
MFLX31200-90
Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Vannes Ball Valves
VWR Item#MFLX31200-90
Your Organization's Price:
Build leakproof fluid handling systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: male x male luer lock, one-way flow pattern
These one-way stopcocks feature an all-black PVDF construction.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: male x male luer lock, one-way flow pattern
More About This Item
These one-way stopcocks feature an all-black PVDF construction.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité