Masterflex® Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Fournisseur: Avantor Fluid Handling
MFLX77301-82EA 786 EUR InStock
MFLX77301-82
Masterflex® Extension Cable for L/S® and I/P® Digital Modular Drives; 9 ft
Câbles
VWR Item#MFLX77301-82
Your Organization's Price:
Increase modular drive flexibility with extended cable lengths.
  • Easily connect to your Masterflex® pump with connectors on both ends
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2,7 m) to increase installation flexibility. Do not extend more than 25 ft.
This extension cable is compatible with L/S® pump models 77301-40, -50, -60, and -70; and with I/P® pump models 07594-00 and -10.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Easily connect to your Masterflex® pump with connectors on both ends
More About This Item
This extension cable is compatible with L/S® pump models 77301-40, -50, -60, and -70; and with I/P® pump models 07594-00 and -10.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité