Mesh Overscreen for Sanitary Strainer 3" dia; 100 x 100 Mesh
Fournisseur: Avantor Fluid Handling
29590-37
MFLX29590-37EA 238 EUR InStock
MFLX29590-37
Mesh Overscreen for Sanitary Strainer 3" dia; 100 x 100 Mesh
Filtres Strainers
VWR Item#MFLX29590-37
Your Organization's Price:
Protect products and expensive process equipment from coarse or fine particulates.
  • Mesh overscreens are available in a variety of mesh sizes to meet your specific application
  • Overscreens are hemmed smooth with no stray wire ends
  • Overscreens fit both in-line and side-inlet sanitary strainer assemblies
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • Mesh overscreens are available in a variety of mesh sizes to meet your specific application
  • Overscreens are hemmed smooth with no stray wire ends
  • Overscreens fit both in-line and side-inlet sanitary strainer assemblies
More About This Item
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité