Precision Digital Process meter, explosion-proof, input loop powered with backlight
Fournisseur: Avantor Fluid Handling
30004-68
MFLX30004-68EA 1020 EUR InStock
MFLX30004-68
Precision Digital Process meter, explosion-proof, input loop powered with backlight
Systèmes de surveillance Process Monitoring
VWR Item#MFLX30004-68
Your Organization's Price:
Four sensors operate as through-glass buttons for easy on-site programming without removing the cover and exposing the electronics.
  • High-contrast backlit LCD is easy to read from a distance and under most lighting conditions
  • 5-digit rate and 7-character total/tag display
  • 32-point, square root, or exponential linearization
  • 90° rotatable display
  • 5-digit password prevents unauthorized changes to the programmed parameter settings
  • Explosion-proof, IP68, NEMA 4X enclosure
  • FM, CSA, ATEX, IECEx, CE approved
The ProtEX PD6800 is a rugged, explosion-proof process meter designed for demanding applications in hazardous areas or in harsh environmental conditions. The meter derives all of its power from the 4 to 20 mA loop. Easily program using the four SafeTouch™ through-glass buttons without having to remove the cover or use the four mechanical buttons. Unit features backlit display, min/max readings, password protection, and alarm indication. Die-cast aluminum enclosure has two ¾" threaded conduit holes and integrated pipe or wall mounting flanges.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
CE Compliant 
Key Features
  • High-contrast backlit LCD is easy to read from a distance and under most lighting conditions
  • 5-digit rate and 7-character total/tag display
  • 32-point, square root, or exponential linearization
  • 90° rotatable display
  • 5-digit password prevents unauthorized changes to the programmed parameter settings
  • Explosion-proof, IP68, NEMA 4X enclosure
  • FM, CSA, ATEX, IECEx, CE approved
More About This Item
The ProtEX PD6800 is a rugged, explosion-proof process meter designed for demanding applications in hazardous areas or in harsh environmental conditions. The meter derives all of its power from the 4 to 20 mA loop. Easily program using the four SafeTouch™ through-glass buttons without having to remove the cover or use the four mechanical buttons. Unit features backlit display, min/max readings, password protection, and alarm indication. Die-cast aluminum enclosure has two ¾" threaded conduit holes and integrated pipe or wall mounting flanges.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité