Masterflex® Stepping-Motor Proportioning Valve, Al; 3/4"NPT(F), 1.5-10 VDC
Fournisseur: Avantor Fluid Handling
98651-08
MFLX98651-08EA 904 EUR InStock
MFLX98651-08
Masterflex® Stepping-Motor Proportioning Valve, Al; 3/4"NPT(F), 1.5-10 VDC
Vannes Solenoid Valves
VWR Item#MFLX98651-08
Your Organization's Price:
Resist pressures up to 500 psi!
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
More About This Item
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité