Dow Corning Transfter Tubing, Pharm-65 Platinum-Cured Silicone, 3/8" ID x 5/8" OD; 50 Ft
Fournisseur: Avantor Fluid Handling
96107-09
MFLX96107-09EA 851 EUR InStock
MFLX96107-09
Dow Corning Transfter Tubing, Pharm-65 Platinum-Cured Silicone, 3/8" ID x 5/8" OD; 50 Ft
Tuyaux Transfer Tubing
VWR Item#MFLX96107-09
Your Organization's Price:
Platinum-cured silicone tubing with a very low extractable profile.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
Certification: Elastomer is FDA compliant and meets USP Class VI requirements. Meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Tubing is manufactured according to Bulk Pharmaceutical (BP) and current GMP. Temperature range: –58 to 482 °F (–50 to 250 °C).
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
CE certificate 
Key Features
  • Global market leader in range of formulations for most extensive chemical compatibility
More About This Item
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
Certification: Elastomer is FDA compliant and meets USP Class VI requirements. Meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Tubing is manufactured according to Bulk Pharmaceutical (BP) and current GMP. Temperature range: –58 to 482 °F (–50 to 250 °C).
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité