Masterflex® Sanitary Tri-Clamp Screen Gasket, Tuf-Flex®, 3/4" Tri-Clamp; 10/PK
Fournisseur: Avantor Fluid Handling
MFLX30547-52EA 234 EUR InStock
MFLX30547-52
Masterflex® Sanitary Tri-Clamp Screen Gasket, Tuf-Flex®, 3/4" Tri-Clamp; 10/PK
Scellés Joints
VWR Item#MFLX30547-52
Your Organization's Price:
Ensure a better process seal with outstanding chemical resistance.
  • The chemical resistance of PTFE and the flexibility of an elastomer
  • Designed for CIP/SIP conditions
The Tuf-Flex gasket grafts a PTFE envelope to an elastomer core. Unlike pure PTFE gaskets, or even other sheathed gaskets, Tuf-Flex will not creep or cold flow over time and therefore maintains the integrity of its seal in your process line. The core gives the gasket the mechanical properties of an elastomer, including memory and flexibility. Designed to meet the stringent requirements of biopharmaceutical, ultra-pure water, WFI (water for injection), and food and beverage processing applications. A USP Class VI certification; meets USDA and 3A sanitary standards. Material is free of animal-derived ingredients.
Other Products In this Family
  See all Items in this Product Family
SPÉCIFICATIONS
Key Features
  • The chemical resistance of PTFE and the flexibility of an elastomer
  • Designed for CIP/SIP conditions
More About This Item
The Tuf-Flex gasket grafts a PTFE envelope to an elastomer core. Unlike pure PTFE gaskets, or even other sheathed gaskets, Tuf-Flex will not creep or cold flow over time and therefore maintains the integrity of its seal in your process line. The core gives the gasket the mechanical properties of an elastomer, including memory and flexibility. Designed to meet the stringent requirements of biopharmaceutical, ultra-pure water, WFI (water for injection), and food and beverage processing applications. A USP Class VI certification; meets USDA and 3A sanitary standards. Material is free of animal-derived ingredients.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité