Would you like to log in again?
VWR® SMART5, Gel Documentation System
Supplier: VWR Collection
730-0496EA 9140 EUR InStock
730-0496
VWR® SMART5, Gel Documentation System
Photodocumentation Systems
SMART5 is a new generation of low cost, integrated imaging system for DNA and protein analysis, also gel documentation.
  • Compact workstation
  • Complete with built-in processor, touch screen and SMART5 software
  • Motor-driven zoom lens
  • 5 MP camera
  • White, UV and blue lighting options
  • Complete networking connectivity (optional)
SMART5 is the ultimate in compact gel documentation workstations. Using a choice of UV, blue or white lighting options, you can capture and edit images of fluorescent ethidium bromide, SYBR® Safe or GelGreen™ DNA gels and protein gelsMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Documentation
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialité..