Would you like to log in again?
Samplers, pigment lance
Supplier: BURKLE
5369-0300 5369-0900 5369-0610 5369-0910 5369-0600 5369-0310
231-0480EA 396 EUR InStock
231-0480 231-0484 231-0483 231-0485 231-0482 231-0481
Samplers, pigment lance
Samplers Solid Samplers
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
  • Half-open flute, 50%, for highly sticky products
  • Quarter-open flute, 25%, for less sticky products
  • Stainless steel V4A (1.4404)
Order Now
ORDER
Documentation
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty