B24303.06
B24303.14
B24303.22
B24303.06EA
31.9
EUR
InStock
B24303.06
B24303.14
B24303.22
Benzhydrylamine 97%
Benzhydrylamine
Purity: 97%
CAS No.: 91-00-9
Molecular Formula: C13H13N
Formula Weight: 183.25
Flash Point: 110°(230°F)
Refractive Index: 1.5950
Melting Point: 11-13°
Boiling Point: 294-296°
Solubility: Slightly soluble in water
Beilstein Reference: 776434
Merck Reference: 14,1076
UN No.: 2735
MDL No.: MFCD00008059