HPLC columns, GlycanPac™ AXR-1
Fournisseur: Thermo Fisher Scientific
GlycanPac™
555-7350EA 234 EUR InStock
555-7350 555-7339 555-7334 555-7335 555-7341 555-7352 555-7340 555-7351
HPLC columns, GlycanPac™ AXR-1
Colonnes pour chromatographie
The GlycanPac™ AXR-1 column uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments. Designed and tested for HPLC and UHPLC methods using either fluorescence or MS detection applications. Ultra-high resolution and selectivity of glycans are attributed to the GlycanPac™ AXR-1 column's innovative mixed-mode surface chemistry, which combines:
  • WAX functionality for retention of negatively charged glycans, with separation in order of increasing charge
  • Reversed-phase functionality for same-charge glycans, with separation according to isomerism and size
Particle type: high purity, spherical, porous silica
Bonded phase: mixed-mode: weak anion exchange and reversed phase
pH range: 2,0 to 8,0
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité