Latex sleeves for dry boxes, with removable hand, SP Bel-Art
Fournisseur: Bel-Art Products, a part of SP
BELAH500250309EA 1190 EUR InStock
BELAH500250309 BELAH500250310
Latex sleeves for dry boxes, with removable hand, SP Bel-Art
Gants
These sleeves with removable hands are made from natural latex. They fit 20 cm (8") circular ports and 18×25 cm (7×10") oval ports.
  • Bellows type
  • Sleeve stays fastened to the box when the glove is disconnected from the sleeve
  • Gloves sizes are interchangeable and virtually any standard laboratory, surgical or disposable glove can be employed
  • An O-ring seal that mates with the glove box wall provides watertight performance for the glove and sleeve unit
  • Cool air circulates through the bellows, allowing comfortable operation
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité