Albumin and IgG depletion columns, HiTrap™
Fournisseur: Cytiva
HiTrap™
28-9466-03EA 1064 EUR InStock
28-9466-03
Albumin and IgG depletion columns, HiTrap™
Colonnes pour chromatographie
HiTrap Albumin and IgG Depletion is a prepacked columns for the depletion of albumin and IgG from human serum and plasma. The columns are prepacked with high performance SepharoseTM based resins with an affinity for human serum albumin (HSA) and IgG.
  • HiTrap Albumin and IgG Depletion is a prepacked column for the depletion of albumin and IgG from human serum or plasma
  • HiTrap Albumin and IgG Depletion column provides rapid and easy processing in a convenient format for handling larger sample volumes (~150 μl) using a liquid chromatography system or manually using a syringe
  • Columns are prepacked with high performance Sepharose based resin with an affinity for human serum albumin (HSA) and IgG
  • High depletion capacity, removes >95% HSA and >90% IgG, with high reproducibility
  • Simple and fast procedure
  • Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins
Albumin and IgG are the most abundant plasma proteins, and they tend to obscure the signals of less abundant proteins. Depletion of the two proteins removes more than 60% of the total protein content in human plasma, allowing a higher load of less abundant proteins to be included in the analysis and detection of more proteins. These columns are members of the Trap platform and address the need for flexible, small-scale preparation of protein samples prior to downstream analyses such as 1-D or 2-D gel electrophoresis and mass spectrometry.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité