CytoScan™-fluoro cytotoxicity assay kit
Fournisseur: G-Biosciences
786-211EA 946 EUR InStock
786-211
CytoScan™-fluoro cytotoxicity assay kit
Tests Tests cellulaires
The CytoScan™-fluoro Cytotoxicity assay is based on the quantification of cellular lactate dehydrogenase (LDH) released when cells are damaged or under stress. Lactate dehydrogenase released into the culture medium is measured with a diaphorase coupled enzymatic assay that results in the conversion of a non-fluorescent compound (resazurin) to a fluorescent compound (resorufin), which is measured using a fluorometer. The assay detects even low level damage to cell membrane not detected with other methods.
  • Fluorometric assay
  • Quantitatively measures LDH release
  • Assays performed directly in cell culture wells
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening
The kit does not damage healthy cells and the assay can be performed directly in the cell culture wells containing a mixed population of viable and damaged cells. The kit is supplied with substrate mix, assay buffer, and stop solution.
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité