Phosphatase assay kit
Fournisseur: G-Biosciences
786-453EA 836 EUR InStock
786-453
Phosphatase assay kit
Tests Tests enzymatiques
The Phosphatase Assay Kit is designed to measure the activity of phosphatases in biological samples and to screen for agonists and inhibitors of phosphatases. The Phosphatase Assay Kit uses para-nitrophenyl phosphate (pNPP), a chromogenic substrate for most phosphatases, including alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases.
  • Colorimetric, pNPP based assay
  • Measure phosphatase activity in biological samples
  • Screen for phosphatase agonists and inhibitors
  • For the quantification of phosphatase activity
  • To screen for agonists and inhibitors of phosphatases
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité