Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD274 Rabbit Polyclonal Antibody

Fournisseur: Boster
BSBTPA1851EA 0 EUR InStock
BSBTPA1851
Anti-CD274 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Programmed cell death 1 ligand 1(CD274) detection. Tested with WB in Human;Mouse;Rat.

CD274(CD274 molecule), also known as PDCD1L1, PDL1, B7H1, CD274 molecule, cluster of differentiation(CD274) or B7 homolog 1(B7-H1), Programmed cell death 1 ligand 1, is a protein that in humans is encoded by the CD274 gene. Levels of B7H1 correlated with PTEN loss in glioblastoma specimens, and tumor-specific T cells lysed human glioma targets expressing wildtype PTEN more effectively than those expressing mutant PTEN. Sequence analysis predicted that it has 290-amino acid. immunoresistance in glioma is related to loss of the tumor suppressor PTEN and is mediated in part by B7H1.Programmed death ligand 1(PD-L1) is a 40kDa type 1 transmembrane protein that has been speculated to play a major role in suppressing the immune system during particular events such as pregnancy, tissue allografts, autoimmune disease and other disease states such as hepatitis. Using microarray analysis and flow cytometry, Barber et al.(2006) found that Pd1 was highly upregulated by functionally exhausted CD8 T cells from mice infected with a lymphocytic choriomeningitis virus(LCMV) strain causing chronic infection, but not by functional memory CD8 T cells from mice infected with an LCMV strain causing acute infection. 

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: CD274
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité