Anti-PINK1 Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
BC100-494 BC100-494SS
NOVUBC100-494EA 738 EUR InStock
NOVUBC100-494 NOVUBC100-494SS
Anti-PINK1 Rabbit Polyclonal Antibody
Anticorps
The PINK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PINK1. This antibody reacts with human, mouse, rat, drosophila. The PINK1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Peptide ELISA.
There are 3 scientific publications using this antibody.
Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Drosophila
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité