Safety trimming blades, FEATHER®
Fournisseur: PFM Medical GmbH
206600260_U 206600130 206600000 02.06600260
FEAT206600260_UEA 147 EUR InStock
FEAT206600260_U FEAT206600130 FEAT206600000 FEAT02.06600260
Safety trimming blades, FEATHER®
Microtomes et groupes de microtomes Lames de microtomes
These trimming blades in conjunction with the handle F-1326P allow fast and easy blade change thus providing users with increased safety and comfort in their daily laboratory work. With back metal, blade can be attached/detached without touching blade directly.
  • Individually wrapped blade holders prevent direct contact with the trimming blades thus protecting against injury and infection
  • Fast and easy blade change
  • Handle is particularly easy to grip due to its ergonomical design
  • Handle is heat resistant, easy to clean and autoclavable
  • Individually packed
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité