Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-TACR1 Rabbit Polyclonal Antibody

Fournisseur: Boster
BSBTPB9446EA 579 EUR InStock
BSBTPB9446
Anti-TACR1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Substance-P receptor(TACR1) detection. Tested with WB in Human;Rat.

The tachykinin receptor 1 (TACR1), also known as neurokinin 1 receptor (NK1R) or substance P receptor (SPR), is a G protein coupled receptor found in the central nervous system and peripheral nervous system. The endogenous ligand for this receptor is Substance P, although it has some affinity for other tachykinins. The protein is the product of the TACR1 gene. Tachykinin receptor 1 consists of 407 amino acid residues, and it has a molecular weight of 58.000. Tachykinin receptor 1, as well as the other tachykinin receptors, is made of seven hydrophobic transmembrane (TM) domains with three extracellular and three intracellular loops, an amino-terminus and a cytoplasmic carboxy-terminus. The loops have functional sites, including two cysteines amino acids for a disulfide bridge, Asp-Arg-Tyr, which is responsible for association with arrestin and, Lys/Arg-Lys/Arg-X-X-Lys/Arg, which interacts with G-proteins.The tachykinin receptor 1 can be found in both the central and peripheral nervous system. It is present in neurons, brainstem, vascular endothelial cells, muscle, gastrointestinal tracts, genitourinary tract, pulmonary tissue, thyroid gland and different types of immune cells. The binding of SP to the tachykinin receptor 1 has been associated with the transmission of stress signals and pain, the contraction of smooth muscles and inflammation. Tachykinin receptor 1 antagonists have also been studied in migraine, emesis and psychiatric disorders. In fact, aprepitant has been proved effective in a number of pathophysiological models of anxiety and depression. Other diseases in which the tachykinin receptor 1 system is involved include asthma, rheumatoid arthritis and gastrointestinal disorders.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for TACR1 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TFAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité