Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-VCAN Rabbit Polyclonal Antibody

Fournisseur: Boster
BSBTPB9453EA 550 EUR InStock
BSBTPB9453
Anti-VCAN Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Versican core protein(VCAN) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Versican (VCAN), also known as CSPG2, is a large extracellular matrix proteoglycan that is present in a variety of human tissues. Versican is a large chondroitin sulfate proteoglycan with an apparent molecular mass of more than 1000kDa. The Versican gene is mapped on 5q14.2-q14.3. The human versican gene contains 15 exons spanning more than 90 kb. The distribution of versican by using affinity-purified polyclonal antibodies that recognize the core protein of the prominent versican splice variants V0 and V1. Versican staining was noted in the central and peripheral nervous system, in the basal layer of the epidermis, and on the luminal surface of some glandular epithelia. Biochemical purification of LLC-conditioned medium led to identification of the extracellular matrix proteoglycan versican, which is upregulated in many human tumors including lung cancer, as a macrophage activator that acts through TLR2 and its coreceptors TLR6 and CD14. By activating TLR2:TLR6 complexes and inducing TNF-alpha secretion by myeloid cells, versican strongly enhances LLC metastatic growth.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for Versican is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: VCP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité