Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-IgG Goat Polyclonal Antibody (CF™ 405M)

Fournisseur: Biotium

20373-500ULEA 347 EUR InStock
20373-500UL
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Anticorps
CF™405 antibodies are affinity-purified antibodies labeled with blue fluorescent CF™405S or CF™405M, two members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™405S and CF™405M are two blue fluorescent dyes optimized for the 405nm laser line. CF™405S has excitation/emission maxima at 404/431nm and is spectrally similar to AlexaFluor®405, Cascade Blue®, and DyLight®405. The emission wavelength of CF™405S is centered within the blue detection window of BD flow cytometers, resulting in a higher signal compared to AlexaFluor® 405. CF™405M has excitation/emission maxima at 408/452nm, and is spectrally similar to BD Horizon™ V450, eFluor®450, and Pacific Blue®. CF™405M is significantly more photostable than Pacific Blue®, and has less spillover in the green channel than other spectrally similar dyes.

CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor®, Cascade Blue®, and Pacific Blue® are registered trademarks of Molecular Probes, Inc. BD Horizon™ is a trademark of BD Biosciences. DyLight® is a registered trademark of Pierce Biotechnology.

Informations de commande: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF405M
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité