Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-BDNF Chicken Polyclonal Antibody

Fournisseur: Biosensis
BSENC-1517-500EA 312 EUR InStock
BSENC-1517-500
Anti-BDNF Chicken Polyclonal Antibody
Anticorps
BDNF belongs to the neurotrophin family and regulates the survival and differentiation of neurons during development. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. FUNCTION: Promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. POst translation modification: Converted into mature BDNF by plasmin (PLG). SIMILARITY: Belongs to the NGF-beta family.

The antibody specifically reacts with BDNF protein. The antibody has been successfully tested in ELISA. The antibody has not been tested in western blot or IHC. BDNF is highly conserved in human, mouse, and rat, as well as other mammalian species so cross reactivity to BDNF from other mammals is expected.
Application Information:
ELISA. Suggested dilution at 1:500 to 1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human BDNF peptides consisting of amino acids 73-87 and 194-208.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité