Anti-NME1 Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTRP1090EA 550 EUR InStock
BSBTRP1090
Anti-NME1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Nucleoside diphosphate kinase A(NME1) detection. Tested with WB in Human;Mouse;Rat.
NME1(NME/NM23 nucleoside diphosphate kinase 1), also called non-metastatic cells 1, protein (NM23A) expressed in, NM23, NM23-H1, NDPKA, GAAD or AWD, is an enzyme that in humans is encoded by the NME1 gene. The promoters of the mouse and human NME1 genes, like those of other NME genes, contain several binding sites for AP2, NF1, Sp1, LEF1, and response elements to glucocorticoid receptors. The NME1 gene is mapped on 17q21.33. Immunofluorescence microscopy demonstrated colocalization of NME1 in nuclei of B cells expressing EBNA3C. Expression of EBNA3C reversed the ability of NME1 to inhibit migration of BL and breast carcinoma cells. NM23H1 bound SET and was released from inhibition by GZMA cleavage of SET. After GZMA loading or cytotoxic T lymphocyte attack, SET and NM23H1 translocated to the nucleus and SET was degraded, allowing NM23H1 to nick chromosomal DNA. Using a Drosophila model system, Dammai et al. (2003) showed that the Drosophila NME1 homolog, awd, regulates trachea cell motility by modulating FGFR levels through a dynamin -mediated pathway.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité