Anti-FLOT2 Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTPB9623EA 550 EUR InStock
BSBTPB9623
Anti-FLOT2 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Flotillin-2(FLOT2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
FLOT2 (Flotillin 2), also known as ESA1 or M17S1, is a protein that in humans is encoded by the FLOT2 gene. Schroeder et al. (1991) isolated a cDNA for an epidermal surface antigen believed to be involved in epidermal cell adhesion. By analysis of a somatic cell hybrid panel and in situ hybridization using the ESA cDNA, the gene was mapped to 17q11-q12 in the region containing the NF1 gene. Bickel et al. (1997) found that mouse Flot2 consistently copurifies with Flot1 and with caveolin-1 in the purification of caveolin-rich membranes. Using a quantitative proteomic analysis of cultured mouse neuronal stem cells, Li et al. (2012) found that palmitoylation and oligomerization of flotillin-2 was abolished in homozygous Dhhc5 mutant neuronal stem cells. The absolute amount of flotillin-2 was not changed in Dhhc5 mutant neurons.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité