Anti-GJC1 Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTPB9624EA 579 EUR InStock
BSBTPB9624
Anti-GJC1 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Gap junction gamma-1 protein(GJC1) detection. Tested with WB in Human;Rat.
Gap junction gamma-1 protein (GJC1), also known as gap junction alpha-7 protein (GJA7) or connexin 45 (Cx45), is a protein that in humans is encoded by the GJC1 gene. The International Radiation Hybrid Mapping Consortium mapped the GJA7 gene to chromosome 17q21.31. This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité