Anti-HDAC6 Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTPB9628EA 579 EUR InStock
BSBTPB9628
Anti-HDAC6 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Histone deacetylase 6(HDAC6) detection. Tested with WB in Human;Rat.
HDAC6, also called KIAA0901, is a member belongs to class II of the histone deacetylase/acuc/apha family of proteins that is an enzyme that in humans is encoded by the HDAC6 gene. The HDAC6 gene is mapped to chromosome Xp11.23. HDAC6 contains an internal duplication of two catalytic domains which appear to function independently of each other. The protein possesses histone deacetylase activity and represses transcription. HDAC6 functions as a tubulin deacetylase. And it is localized exclusively in the cytoplasm, where it associates with microtubules and localizes with the microtubule motor complex. HDAC6 could bind both polyubiquitinated misfolded proteins and dynein motors, thereby recruiting misfolded protein cargo to dynein motors for transport to aggresomes. Furthermore, expression of HDAC6 was sufficient to rescue degeneration associated with UPS dysfunction in vivo in an autophagy-dependent manner. HDAC6 is a central component of the stress response that regulates SG formation and potentially contributes to control of RNA metabolism and translation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité