Anti-HDAC11 Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTPB9630EA 550 EUR InStock
BSBTPB9630
Anti-HDAC11 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Histone deacetylase 11(HDAC11) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Histone deacetylase 11 is a 39kDa histone deacetylase enzyme that in humans is encoded by the HDAC11 gene on chromosome 3 in humans and chromosome 6 in mice. This gene encodes a class IV histone deacetylase. And the encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants. HDAC11 expression is normally found in brain and testis tissue, but upregulation of HDAC11 expression has also been seen in various cancer cells. In addition, HDAC11 has been shown to be a negative regulator of IL-10 production in antigen presenting cells. It has also been shown that inhibition of HDAC11 results in increased expression of OX40L in Hodgkin lymphoma cells.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité