Synonyms:
Triphenylaminomethane, Tritylamine, Triphenylmethanamine
A13864.03
A13864.06
A13864.14
A13864.22
A13864.03EA
37.8
EUR
InStock
A13864.03
A13864.06
A13864.14
A13864.22
Triphenylmethylamine 98%
Triphenylmethylamine
Purity: 98%
CAS No.: 5824-40-8
Molecular Formula: (C6H5)3CNH2
Formula Weight: 259.35
Melting Point: 100-104°
Boiling Point: 223°/14mm
Beilstein Reference: 2113674
MDL No.: MFCD00008047