A18226.18
A18226.30
A18226.18EA
36.8
EUR
InStock
A18226.18
A18226.30
Dibromofluorescein 95% (teneur en colorant)
Dibromofluorescein
Purity: 95%
CAS No.: 596-03-2
Molecular Formula: C20H10Br2O5
Formula Weight: 490.12
Melting Point: 270-273°
Beilstein Reference: 57189
MDL No.: MFCD00005042