Synonyms:
N,N'-Methylenebisformamide, N,N'-Methylenediformamide
B24116.03
B24116.06
B24116.03EA
28.1
EUR
InStock
B24116.03
B24116.06
Methylenediformamide 97%
Methylenediformamide
Purity: 97%
CAS No.: 6921-98-8
Molecular Formula: C3H6N2O2
Formula Weight: 102.09
Melting Point: 141-143°
Beilstein Reference: 1811705
MDL No.: MFCD00021038