Synonyms:
DL-Phosphinothricin, monoammonium salt
J66186.MD
J66186.03
J66186.MDEA
112
EUR
InStock
J66186.MD
J66186.03
Glufosinate ammonium 95%
Glufosinate ammonium
Purity: 95%
CAS No.: 77182-82-2
Molecular Formula: C5H15N2O4P
Formula Weight: 198.16
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Physical Form: Powder
Appearance: Off-white
Melting Point: 215°
Boiling Point: 519°
Solubility: Soluble in water
Beilstein Reference: 8163399
MDL No.: MFCD00055562
Applications: Interferes with the synthesis of the amino acid glutamine