Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Fournisseur: Biotium
20412-100UL 20412-500UL
20412-100ULEA 161 EUR InStock
20412-100UL 20412-500UL
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
This product is prepared by labeling goat anti-rabbit IgG (H+L) with allophycocyanin (APC). APC belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. APC has absorption maxima at 650 and emission at 660 nm. Because of the high fluorescence quantum yield (~0.68), APC is by far the brightest dye for the 650 nm wavelength region.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité