ACRO104730050EA
96.1
EUR
InStock
ACRO104730050
D-Amygdalin hydrate 95%
D-Amygdalin hydrate
CAS No.: 29883-15-6
MDL No.: MFCD00006598
GHS Symbols: GHS07
Melting Point: 223°C - 226°C
Appearance: White Crystalline powder
Storage Temperature: Ambient
Signal word: Warning