Anti-ZWINT Rabbit Polyclonal Antibody
Fournisseur: Boster
BSBTPB10018EA 550 EUR InStock
BSBTPB10018
Anti-ZWINT Rabbit Polyclonal Antibody
Anticorps
ZW10 interactor (Zwint-1) is a protein that in humans is encoded by the ZWINT gene. This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
1. "Entrez Gene: ZWINT ZW10 interactor".
2. Obuse C, Iwasaki O, Kiyomitsu T, Goshima G, Toyoda Y, Yanagida M (Nov 2004). "A conserved Mis12 centromere complex is linked to heterochromatic HP1 and outer kinetochore protein Zwint-1". Nature Cell Biology 6 (11): 1135–41.
3. van Vlijmen T, Vleugel M, Evers M, Mohammed S, Wulf PS, Heck AJ, Hoogenraad CC, van der Sluijs P (Aug 2008). "A unique residue in rab3c determines the interaction with novel binding protein Zwint-1". FEBS Letters 582 (19): 2838–42.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité