BrightVision Anti-IgG (H+L) (Ultimate kit)
Fournisseur: Immunologic
UBVO500HRPEA 4540 EUR InStock
UBVO500HRP UBVO110HRP S/UBVO-HRP UBVO999HRP
BrightVision Anti-IgG (H+L) (Ultimate kit)
Anticorps
BrightVision immunohistostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, these kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies. These advantages would bring to laboratories the benefit of more accurate result, faster turn-around, less trouble shooting and better costs-saving. The polymer complex will be visualized with the Bright DAB appropriate substrate or chromogen.
  • High-quality secondary antibody and substrate for IHC applications
  • Antigen specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains poly-HRP-anti-mouse/rabbit IgG (ready-to-use) and Bright DAB (substrate + chromogen)
Certifications: Meets CE/IVD standards.
Information de livraison: Shipped with blue ice. Store between 2 to 8 °C.
Attention: DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. DAB can be deactivated with equal parts of Clorox.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité