L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
2,6-Di-tert-butyl-p-crésol, poudre cristalline
Fournisseur: MP Biomedicals
Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
0210116280 0210116290 0210116291
ICNA0210116280EA 23.9 EUR InStock
ICNA0210116280 ICNA0210116290 ICNA0210116291
2,6-Di-tert-butyl-p-crésol, poudre cristalline
2,6-Di-tert-butyl-p-crésol
Butylated hydroxytoluene is a phenolic antioxidant.
Butylated hydroxytoluene (BHT) has been shown to inhibit lipid peroxidation. It causes lung injury and promotes tumors in mice, but this may be due to a metabolite of BHT, 6-tert-butyl-2-[2′-(2′-hydroxymethyl)-propyl]-4-methylphenol. Metabolites of BHT have also been reported to induce DNA strand breaks and internucleosomal DNA fragmentation (a characteristic of apoptosis) in cultured cells. In rats, a single intraperitoneal injection of BHT (60 mg/kg body mass) results in a significant increase in nuclear DNA methyl transferase activity in the liver, kidneys, heart, spleen, brain and lungs. Incubation of alveolar macrophages with BHT significantly reduced the level of TNF-α which may explain the mechanism by which this antioxidant reduces inflammation. Preincubation of aspirin-treated platelets with BHT inhibits the secretion, aggregation, and protein phosphorylation induced by protein kinase C activators. BHT was also found to inhibit the initiation of hepatocarcinogenesis by aflatoxin B1.
Insoluble in water; freely soluble in toluene, soluble in methanol, ethanol (100 mg/ml), isopropanol, methyl ethyl ketone, acetone, Cellosolve, benzene and most other hydrocarbon solvents. More soluble in food oils and fats than butylated hydroxyanisole.
Attention: Research Use Only (RUO).
Formule: C₁₅H₂₄O
MW: 220.35 g/mol
Point d'ébullition: 265
Point d'éclair: 127
Température de stockage: Température ambiante
Numéro MDL: MFCD00011644
Numéro CAS: 128-37-0
Merck Index: 13,01547
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
FTIR Conforms to Standard
Melting Point 68 - 74 °C
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité