Bovine native thrombin (from Plasma)
Fournisseur: MP Biomedicals
ICNA0215416350EA 629 EUR InStock
ICNA0215416350 ICNA0215416310 ICNA0215416301 ICNA0215416305 ICNA0215416380
Bovine native thrombin (from Plasma)
Protéines et Peptides
Thrombin is prepared from fresh bovine plasma by an ion exchange method. Fibrinogen is removed from the prothrombin complex by precipitation. The concentrated prothrombin complex is then activated by beef thromboplastin and calcium. The thrombin solution is clarified by centrifugation and filtration and then lyophilised.
  • Enzyme activity: ≥1500 NIH U/mg protein
  • Enzyme Commission Number: E.C. 3.4.21.5
  • Synonyms: Factor IIa
  • Plasminogen and plasmin not detected
Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.
α-Thrombin is a trypsin-like serine protease involved in a multitude of processes in the human body. Thrombin generation is the result of limited proteolysis of the vitamin K-dependent zymogen prothrombin. Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin which forms the fibrin gel of a hemostatic plug or a pathologic thrombus. In addition, thrombin potentiates the procoagulant process by activating Factors V, VIII, XI and XIII.
Unit definition: Amount required to clot 1 ml of standardised fibrinogen solution in 15 seconds.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité