Bovine serum albumin (BSA), Fraction V
Fournisseur: MP Biomedicals
ICNA08810032EA 173 EUR InStock
ICNA08810032 ICNA08810034 ICNA08810033
Bovine serum albumin (BSA), Fraction V
Protéines et Peptides
Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.It is used to solubilise lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).
  • Protease-free
  • Supplied as crystalline flakes
  • Prior to lyophilisation, the product is filtered through a 0,2 µm filter
  • Soluble in 0,0232, 4% solution, delta A550
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité